

Haematoxylin modified solution Gill III used in nuclear staining, Sigma-Aldrich®
: Merck
:517-28-2
:208-237-3
:C₁₆H₁₄O₆
:MFCD00078111
:302.28 g/mol
:1.07 g/cm3 (20 °C)
:GHS05
:GHS08
:Ambient
:1
- Suitability for microscopy:passes test
- Nuclei:dark blue to dark violet
- Cytoplasm:pink to red
- intercellular substances:pink to red
- Erythrocytes:yellow to orange
- Eosinophilic cytoplasm:pink
Frequently Bought Together