ContactUsLinkComponent
Touch-free soap dispenser, MicroDispenser™
Touch-free soap dispenser, MicroDispenser™
Catalog # MICRICDISP-4
Supplier:  MICRONOVA
undefined
Touch-free soap dispenser, MicroDispenser™
Catalog # MICRICDISP-4
Supplier:  MICRONOVA
Specifications
  • Capacity:
    500 ml
  • Description:
    Touch-free dispenser for Micronova 500 ml pump bottles
  • Pk:
    1
Specifications
About this item
Touch-free soap dispensing unit effectively reduces cross-contamination and waste in the cleanroom and gowning areas.
  • Reduces cross-contamination and waste
  • Activated by an electronic eye
  • Delivers a consistent, premeasured dose of soap, lotion, or sanitiser into the palm
  • Gravity fed with no tubes or pumps
  • No-drip unit may be wall-mounted
Easy delivery system allows dispenser to be use for different soaps or lotions by changing spout. Can be used with 500 ml pump bottles.
Certifications: Complies with OSHA, CDC, and ADA requirements.
Delivery: Operates on four size AA batteries which are included.