Order Entry
Austria
ContactUsLinkComponent
Supelco® Organic Reference Standard, Valeraldehyde
Supelco® Organic Reference Standard, Valeraldehyde
Catalog # 42272-1ML
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Valeraldehyde
Catalog # 42272-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    110-62-3
  • Description:
    Valeraldehyde
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H225 - H317 - H319 - H332 - H335
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00007026
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    Pentanal
  • Melting point:
    –92 °C (lit.)
  • Formula:
    CH₃(CH₂)₃CHO
  • Molecular weight:
    86,13
  • Density:
    0,81 g/ml at 25 °C (lit.)
  • Flash point:
    7 °C
  • Boiling point:
    102...103 °C (lit.)
  • Form:
    Neat

Specifications

About this item

  • GHS signal: GHS02, GHS07
Caution: P210 - P233 - P280 - P303 + P361 + P353 - P304 + P340 + P312 - P305 + P351 + P338