Supelco® Organic Reference Standard, Umckalin
Catalog # 50792-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:43053-62-9
- Description:Umckalin
- Storage conditions:2…8 °C
- Shipping conditions:STD
- MDL No.:MFCD16036238
- M-Clarity quality level:MQ100
- Synonym(s):5-Methoxyscopoletin; 7-Hydroxy-5, 6-dimethoxycoumarin; 7-Hydroxy-5,6-dimethoxy-2H-1-benzopyran-2-one
- Molecular weight:222,19
Specifications
About this item
- GHS signal: GHS07