Order Entry
Austria
ContactUsLinkComponent
Supelco® Organic Reference Standard, Umbelliferone
Supelco® Organic Reference Standard, Umbelliferone
Catalog # 54826-50MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Umbelliferone
Catalog # 54826-50MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Standards:
    Analytical standard
  • CAS No.:
    93-35-6
  • Description:
    Umbelliferone
  • Storage conditions:
    2…8 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00006878
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    7-Hydroxy-2H-1-benzopyran-2-one; 7-Hydroxycoumarin
  • Melting point:
    230 °C (dec.) (lit.)
  • Molecular weight:
    162,14

Specifications

About this item