Order Entry
Austria
ContactUsLinkComponent
Supelco® Organic Reference Standard, Thujone Standard Mixture
Supelco® Organic Reference Standard, Thujone Standard Mixture
Supplier:  Merck
Certificates
Supelco® Organic Reference Standard, Thujone Standard Mixture
Supplier:  Merck

Specifications

  • CAS No.:
    76231-76-0
  • Storage conditions:
    2...8 °C
  • Hazard class description:
    H302
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00135597
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    α,β-Thujone; (−)-1-Isopropyl-4-methylbicyclo[3.1.0]hexan-3-one
  • Formula:
    C₁₀H₁₆O
  • Molecular weight:
    152,23
  • Storage temperature:
    2...8 °C
  • Density:
    0,925 g/ml at 25 °C (lit.)
  • Flash point:
    67 °C - closed cup
  • Boiling point:
    84...86 °C/17 mm Hg (lit.)

Specifications

Frequently Bought Together