Order Entry
Austria
ContactUsLinkComponent
Supelco® Organic Reference Standard, Tetrahydrocurcumin
Supelco® Organic Reference Standard, Tetrahydrocurcumin
Catalog # 50202-10MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Tetrahydrocurcumin
Catalog # 50202-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    36062-04-1
  • Description:
    Tetrahydrocurcumin
  • Storage conditions:
    2...30 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD04152347
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    1,7-Bis(4-hydroxy-3-methoxyphenyl)-3,5-heptanedione
  • Molecular weight:
    372,41
  • Flash point:
    196,2 °C

Specifications

About this item