Order Entry
Austria
ContactUsLinkComponent
Phytochemical standard, Spiraeoside, analytical standard, Supelco®
Phytochemical standard, Spiraeoside, analytical standard, Supelco®
  91802-5MG
 :  Merck
 :  
undefined
Phytochemical standard, Spiraeoside, analytical standard, Supelco®
  91802-5MG
 :  Merck
 :  

 

  • Pk:
    5 mg
  • Standards:
    Analytical standard
  • CAS No.:
    20229-56-5
  • Description:
    Spiraeoside, ≥95,0% (HPLC)
  • Formula:
    C₂₁H₂₀O₁₂
  • Description:
    Spiraeoside
  • Form:
    Neat

 

 

Spiraeoside is a naturally occurring flavonoid, that can extracted from the flowers of Filipendula ulmaria.

 : Bottomless glass bottle. Contents are inside inserted fused cone.