Order Entry
Austria
ContactUsLinkComponent
Organic reference standard, Solvent Yellow 124, analytical standard, Supelco®
Organic reference standard, Solvent Yellow 124, analytical standard, Supelco®
Supplier:  Merck
Certificates
undefined
Organic reference standard, Solvent Yellow 124, analytical standard, Supelco®
Supplier:  Merck

Specifications

  • CAS No.:
    34432-92-3
  • Formula:
    C₂₂H₃₁N₃O₂
  • Storage temperature:
    2...8 °C
  • Description:
    Solvent Yellow 124
  • M-Clarity quality level:
    MQ100

Specifications

Frequently Bought Together