Order Entry
Austria
ContactUsLinkComponent
Phytochemical standard, Silybin B, analytical standard, Supelco®
Catalog # 59527-5MG
Supplier:  Merck
CAS Number:  
undefined
Phytochemical standard, Silybin B, analytical standard, Supelco®
Catalog # 59527-5MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Standards:
    Analytical standard
  • CAS No.:
    142797-34-0
  • Description:
    Silybin B
  • Formula:
    C₂₅H₂₂O₁₀
  • Description:
    Silybin B
  • Form:
    Neat

Specifications

About this item

Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.