Order Entry
Austria
ContactUsLinkComponent
Organic reference standard, Sclareol, analytical standard, Supelco®
Organic reference standard, Sclareol, analytical standard, Supelco®
Catalog # 49944-100MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Sclareol, analytical standard, Supelco®
Catalog # 49944-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    515-03-7
  • Description:
    Sclareol
  • Melting point:
    95 - 100 °C
  • Formula:
    C₂₀H₃₆O₂
  • Description:
    Sclareol
  • Freezing point:
    218 - 220 °C/19 mmHg
  • Form:
    Neat

Specifications

About this item