Supelco® Organic Reference Standard, Salidroside
43866-25MG
: Merck
:
- Pk:25 mg
- Standards:Analytical standard
- CAS No.:10338-51-9
- Description:Salidroside
- Storage conditions:2...30 °C
- Hazard class description:H319
- Shipping conditions:STD
- MDL No.:MFCD00210553
- M-Clarity quality level:MQ100
- Synonym(s):2-(4-Hydroxyphenyl)ethyl β−ᴅ-glucopyranoside; p-Hydroxyphenethyl glucopyranoside; Rhodioloside; Rhodioloside; Rhodosin; Tyrosol a-(β-ᴅ-glucopyranoside)
- Formula:C₁₄H₂₀O₇
- Molecular weight:300,30
- Flash point:Not applicable
- Form:Neat
- GHS signal: GHS07
: Bottomless glass bottle. Contents are inside inserted fused cone.
: P264 - P280 - P305 + P351 + P338 - P337 + P313