Order Entry
Austria
ContactUsLinkComponent
 
Organic reference standard, Rebaudioside D, analytical standard, Supelco®
Catalog # 19189-10MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Rebaudioside D, analytical standard, Supelco®
Catalog # 19189-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    63279-13-0
  • Description:
    Rebaudioside D, ≥90,0% (HPLC)
  • Formula:
    C₅₀H₈₀O₂₈
  • Description:
    Rebaudioside D
  • Form:
    Neat

Specifications

About this item

Rebaudioside D, a glycoside of the diterpene steviol is a minor constituent found in leaves of Stevia rebaudiana. It is considered non-caloric and is about 200 to 220 times sweeter than sucrose.

It may be used as internal standard to optimise the desorption electrospray ionisation (DESI) mass spectrometry during the analysis of Stevia leaves for diterpene glycosides.