Order Entry
Austria
ContactUsLinkComponent
Phytochemical standard, Pseudohypericin, analytical standard, Supelco®
Phytochemical standard, Pseudohypericin, analytical standard, Supelco®
Catalog # 51593-5MG
Supplier:  Merck
CAS Number:  
undefined
Phytochemical standard, Pseudohypericin, analytical standard, Supelco®
Catalog # 51593-5MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Standards:
    Analytical standard
  • CAS No.:
    55954-61-5
  • Description:
    Pseudohypericin, ≥98,0% (HPLC)
  • Formula:
    C₃₀H₁₆O₉
  • Description:
    Pseudohypericin

Specifications

About this item

One of the major bioactive constituents of St. John′s wort (Hypericum perforatum). Implicated in the antidepressant efficacy of St. John′s wort.