Supelco® Organic Reference Standard, Protein Standard Set
Catalog # P5494-1SET
Supplier: Merck
CAS Number:
Specifications
- Pk:1 SET
- Standards:Analytical standard
- Description:Protein standard set
- Storage conditions:2…8 °C
- Shipping conditions:STD
- M-Clarity quality level:MQ200