Order Entry
Austria
ContactUsLinkComponent
Organic reference standard, Propiophenone, analytical standard, Supelco®
Organic reference standard, Propiophenone, analytical standard, Supelco®
Catalog # 61074-1ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Propiophenone, analytical standard, Supelco®
Catalog # 61074-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    93-55-0
  • Description:
    Propiophenone, ≥97,0% (GC)
  • Seq:
    01
  • Melting point:
    17…19 °C
  • Formula:
    C₉H₁₀O
  • Description:
    Propiophenone
  • Flash point:
    99 °C
  • Boiling point:
    218 °C
  • Form:
    Neat

Specifications

About this item