Organic reference standard, Pectin, for use with total dietary fiber control kit, TDF-C10, Supelco®
Catalog # P7536-1G
Supplier: Merck
CAS Number:
Specifications
- Pk:1 g
- CAS No.:9000-69-5
- Description:Pectin
- Environmentally Preferable:
- Storage temperature:2...8 °C
- Description:Pectin
- Form:Lyophilised powder
Specifications
About this item
Packaging: 1 g in poly bottle.