Organic reference standard, N-Pivaloylglycine, analytical standard, Supelco®
: Merck
Pivaloyl glycine is an acylglycine. Acylglycines possess a common amidoacetic acid moiety and are normally minor metabolites of fatty acids. Elevated levels of certain acylglycines appear in the urine and blood of patients with various fatty acid oxidation disorders.
- CAS No.:23891-96-5
- Formula:C₇H₁₃NO₃
- Storage temperature:2...8 °C
- Description:N-Pivaloylglycine
Frequently Bought Together