Organic reference standard, Isovaleraldehyde, analytical standard, Supelco®
Catalog # 61848-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:590-86-3
- Description:Isovaleraldehyde, ≥97,0% (GC)
- Formula:(CH₃)₂CHCH₂CHO
- Storage temperature:2...8 °C
- Description:Isovaleraldehyde
- Boiling point:90 °C
- Form:Neat