Order Entry
Austria
ContactUsLinkComponent
 
Phytochemical standard, Isosilybin B, analytical standard, Supelco®
  95684-5MG
 :  Merck
 :  
undefined
Phytochemical standard, Isosilybin B, analytical standard, Supelco®
  95684-5MG
 :  Merck
 :  

 

  • Pk:
    5 mg
  • Standards:
    Analytical standard
  • CAS No.:
    142796-22-3
  • Description:
    Isosilybin B, ≥80,0% (HPLC)
  • Formula:
    C₂₅H₂₂O₁₀
  • Description:
    Isosilybin B
  • Form:
    Neat

 

 

 : Bottomless glass bottle. Contents are inside inserted fused cone.