Supelco® Organic Reference Standard, Isoliquiritigenin
Catalog # 38986-50MG
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- Standards:Analytical standard
- CAS No.:961-29-5
- Description:Isoliquiritigenin
- Storage conditions:2...8 °C
- Shipping conditions:STD
- MDL No.:MFCD00075907
- M-Clarity quality level:MQ100
- Synonym(s):(E)-1-(2,4-Dihydroxyphenyl)-3-(4-hydroxyphenyl)-2-propen-1-one; 4,2′,4′-Trihydroxychalcone
- Formula:C₁₅H₁₂O₄
- Molecular weight:256,25
- Storage temperature:2...8 °C
- Flash point:Not applicable