Order Entry
Austria
ContactUsLinkComponent
Organic reference standard, Hydroxycitronellal, analytical standard, Supelco®
Organic reference standard, Hydroxycitronellal, analytical standard, Supelco®
Catalog # 66010-1ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Hydroxycitronellal, analytical standard, Supelco®
Catalog # 66010-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    107-75-5
  • Description:
    Hydroxycitronellal, ≥95,0% (GC)
  • Formula:
    C₁₀H₂₀O₂
  • Storage temperature:
    2...8 °C
  • Description:
    Hydroxycitronellal
  • Boiling point:
    257 °C
  • Form:
    Neat

Specifications

About this item

Hydroxycitronellal is a synthetic fragrance material used in perfumes. It can be prepared from citronellal either by hydration of citronellal bisulfite or by hydrolysis with sulphuric acid and alkali.