Order Entry
Austria
ContactUsLinkComponent
Supelco® Organic Reference Standard, HPLC Peptide Standard Mixture
Supelco® Organic Reference Standard, HPLC Peptide Standard Mixture
Artikelnr. H2016-1VL
Lieferant:  Merck
CAS-Nummer:  
undefined
Supelco® Organic Reference Standard, HPLC Peptide Standard Mixture
Artikelnr. H2016-1VL
Lieferant:  Merck
CAS-Nummer:  

Spezifikationen

  • Pk:
    1
  • Standards:
    Analytical standard
  • Description:
    HPLC peptide standard mixture
  • Storage conditions:
    –20 °C
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ200

Spezifikationen

Über diesen Artikel