Supelco® Organic Reference Standard, Hexanal
Hexanal, Analytical standard, Neat, 1 mL
18109-1ML
: Merck
- GHS signal: GHS02, GHS07
: P210 - P233 - P240 - P241 - P303 + P361 + P353 - P305 + P351 + P338
- Pk:1 mL
- Standards:Analytical standard
- Description:Hexanal
- Form:Neat
- Boiling point:130...131 °C (lit.)
- Hazard class description:H226 - H315 - H319
- MDL No.:MFCD00007027
- Density:0,815 g/ml at 25 °C (lit.)
- Synonym(s):Aldehyde C₆; Caproaldehyde; Hexyl aldehyde
- Molecular weight:100,16
- CAS No.:66-25-1
- Cat. No.:18109-1ML
- M-Clarity quality level:MQ100
- Formula:CH₃(CH₂)₄CHO
- Storage conditions:2...30 °C
- Flash point:25 °C - closed cup
- Shipping conditions:STD
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



