Organic reference standard, Heptaldehyde, analytical standard, Supelco®
61696-1ML
: Merck
:
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:111-71-7
- Description:Heptaldehyde, ≥97,0% (GC)
- Melting point:−43 °C
- Formula:CH₃(CH₂)₅CHO
- Storage temperature:2...8 °C
- Description:Heptaldehyde
- Boiling point:153 °C
- Form:Neat