Order Entry
Austria
ContactUsLinkComponent
Supelco® Organic Reference Standard, Genistein
Supelco® Organic Reference Standard, Genistein
Catalog # 92136-10MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Genistein
Catalog # 92136-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    446-72-0
  • Description:
    Genistein
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H302
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00016952
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    4′,5,7-Trihydroxyisoflavone; 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-1-benzopyran-4-one
  • Molecular weight:
    270,24

Specifications

About this item

  • GHS signal: GHS07
Caution: P264 - P270 - P301 + P312 - P501