Supelco® Organic Reference Standard, γ-Tocotrienol
Catalog # 49634-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:14101-61-2
- Description:γ-Tocotrienol
- Storage conditions:–20 °C
- Shipping conditions:DICE
- MDL No.:MFCD05864856
- M-Clarity quality level:MQ100
- Synonym(s):(R)-γ-Tocotrienol; [R-(E,E)]-3,4-Dihydro-2,7,8-trimethyl-2-(4,8,12-trimethyl-3,7,11-tridecatrienyl)-2H-1-benzopyran-6-ol
- Molecular weight:410,63