Order Entry
Austria
ContactUsLinkComponent
Organic reference standard, Fumonisin B2-13C34 solution, analytical standard, Supelco®
Organic reference standard, Fumonisin B2-13C34 solution, analytical standard, Supelco®
Catalog # 32915-1ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Fumonisin B2-13C34 solution, analytical standard, Supelco®
Catalog # 32915-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    10 µg/ml in acetonitrile, water
  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    1217481-36-1
  • Description:
    Fumonisin B₂-¹³C₃₄
    solution
  • M-Clarity quality level:
    MQ100
  • Formula:
    ¹³C₃₄H₅₉NO₁₄
  • Storage temperature:
    −20°C
  • Description:
    Fumonisin B₂-¹³C₃₄
    solution

Specifications

About this item