Order Entry
Austria
ContactUsLinkComponent
Organic reference standard, Cadaverine, analytical standard, Supelco®
Organic reference standard, Cadaverine, analytical standard, Supelco®
  52063-1ML
 :  Merck
 :  
undefined
Organic reference standard, Cadaverine, analytical standard, Supelco®
  52063-1ML
 :  Merck
 :  

 

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    462-94-2
  • Description:
    Cadaverine, ≥96,5% (GC), 96,5 - 103,5% (T)
  • Melting point:
    14...16 °C (lit.)
  • Formula:
     C₅H₁₄N₂
  • Description:
    Cadaverine
  • Boiling point:
    178...180 °C (lit.)
  • Form:
    Neat