Organic reference standard, Bisphenol E, analytical standard, Supelco®
Catalog # 04487-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:2081-08-5
- Description:Bisphenol E, ≥98,0% (HPLC)
- Melting point:123...127 °C (lit.)
- Formula:C₁₄H₁₄O₂
- Description:Bisphenol E
Specifications
About this item
Bisphenol E may be used as a reference standard in the determination of bisphenol E in beverages using on-line solid-phase extraction with fast liquid chromatography coupled with tandem mass spectrometry (on-line SPE fast LC-MS/MS).