Order Entry
Austria
ContactUsLinkComponent
Supelco® Organic Reference Standard, Amino Acid Standards for Protein Hydrolysates
Supelco® Organic Reference Standard, Amino Acid Standards for Protein Hydrolysates
Catalog # A9781-5ML
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, Amino Acid Standards for Protein Hydrolysates
Catalog # A9781-5ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    5 mL
  • Standards:
    Analytical standard
  • Description:
    Amino acid standards for protein hydrolysates
  • Storage conditions:
    2…8 °C
  • Shipping conditions:
    STD

Specifications

About this item