Order Entry
Austria
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, Amino Acid Standards for Fluorescence Detection
 :  Merck
 
Supelco® Organic Reference Standard, Amino Acid Standards for Fluorescence Detection
 :  Merck

 

  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H290
  • Shipping conditions:
    STD

 

Frequently Bought Together