Supelco® Organic Reference Standard, Amino Acid Standard
Supplier: Merck
Certificates
About this item
Caution: P234 - P390
Specifications
- Storage conditions:2…8 °C
- Hazard class description:H290
- Shipping conditions:STD
Specifications
Frequently Bought Together