Organic reference standard, Amaranth, analytical standard, Supelco®
Catalog # 87612-25MG
Supplier: Merck
CAS Number:
Specifications
- Pk:25 mg
- Standards:Analytical standard
- CAS No.:915-67-3
- Description:Amaranth, ≥98,0% (HPLC)
- Formula:C₂₀H₁₁N₂Na₃O₁₀S₃
- Description:Amaranth
- Form:Neat