Order Entry
Austria
ContactUsLinkComponent
Organic reference standard, Alternariol, analytical standard, Supelco®
Organic reference standard, Alternariol, analytical standard, Supelco®
Catalog # 35758-0.1MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Alternariol, analytical standard, Supelco®
Catalog # 35758-0.1MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    0,1 mg
  • Standards:
    Analytical standard
  • CAS No.:
    641-38-3
  • Description:
    Alternariol
  • Formula:
    C₁₄H₁₀O₅
  • Storage temperature:
    –20 °C
  • Description:
    Alternariol
  • Form:
    Neat

Specifications

About this item

Reconstitution: Dried down, 100 μg/ml after reconstitution with 1 ml of water.