Organic reference standard, 4'-Methoxyacetophenone, analytical standard, Supelco®
Catalog # 52222-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:100-06-1
- Description:4'-Methoxyacetophenone, ≥99,0% (GC)
- Melting point:36...38 °C (lit.)
- Formula:C₉H₁₀O₂
- Description:4'-Methoxyacetophenone
- Flash point:138 °C
- Boiling point:152...154 °C/26 mm Hg (lit.)
- Form:Neat