Organic reference standard, 3-Methoxyphenol, analytical standard, Supelco®
Catalog # 44777-1ML
Supplier: Merck
CAS Number:
Specifications
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:150-19-6
- Description:3-Methoxyphenol, ≥96,0% (GC)
- Formula:C₇H₈O₂
- Description:3-Methoxyphenol
- Boiling point:113...115 °C/5 mm Hg (lit.)