Order Entry
Austria
ContactUsLinkComponent
Organic reference standard, 2-Methylindole, analytical standard, Supelco®
Organic reference standard, 2-Methylindole, analytical standard, Supelco®
Catalog # 41069-100MG
Supplier:  Merck
CAS Number:  
Organic reference standard, 2-Methylindole, analytical standard, Supelco®
Catalog # 41069-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    95-20-5
  • Description:
    2-Methylindole, ≥98,0% (HPLC)
  • Melting point:
    57...59 °C (lit.)
  • Formula:
    C₉H₉N
  • Description:
    2-Methylindole
  • Boiling point:
    273 °C (lit.)
  • Form:
    Neat

Specifications

About this item

Regioselective synthesis of oxopyrrolidine analogs via iodine-catalysed Markovnikov addition reaction: Friedel-crafts alkylation reactions, preparation of tryptophan dioxygenase inhibitors, pyridyl-ethenyl-indoles as potential anticancer immunomodulators, preparation of plant-growth inhibitors, michael addition reactions, synthesis of cyclooxygenase-1 (COX-1)/cyclooxygenase-2 (COX-2) inhibitors.