Order Entry
Austria
ContactUsLinkComponent
Organic reference standard, 2-Ethylpyridine, analytical standard, Supelco®
Organic reference standard, 2-Ethylpyridine, analytical standard, Supelco®
Catalog # 68558-1ML
Supplier:  Merck
CAS Number:  
Organic reference standard, 2-Ethylpyridine, analytical standard, Supelco®
Catalog # 68558-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    100-71-0
  • Description:
    2-Ethylpyridine, ≥97,0% (GC)
  • Formula:
    C₇H₉N
  • Description:
    2-Ethylpyridine
  • Boiling point:
    149 °C (lit.)

Specifications

About this item