Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Supelco® Organic Reference Standard, 2-Ethylfuran, Analytical Standard
Supelco® Organic Reference Standard, 2-Ethylfuran, Analytical Standard
Supelco® Organic Reference Standard, 2-Ethylfuran, Analytical Standard
Catalog # 69530-1ML
Supplier:
Merck
CAS Number:
Expand image
Supelco® Organic Reference Standard, 2-Ethylfuran, Analytical Standard
Catalog # 69530-1ML
Supplier:
Merck
CAS Number:
Specifications
Pk:
1 mL
Standards:
Analytical standard
Description:
2-Ethylfuran
Seq:
01
Specifications
About this item
Product Details
Product Details