Supelco® Organic Reference Standard, (+)-Borneol
68878-10MG
: Merck
:
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:464-43-7
- Description:(+)-Borneol
- Storage conditions:2…8 °C
- Hazard class description:H228
- Shipping conditions:STD
- MDL No.:MFCD00066427
- M-Clarity quality level:MQ100
- Synonym(s):endo-(1R)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-ol
- Melting point:206 - 209 °C (lit.)
- Molecular weight:154,25
- Flash point:65 °C - Closed cup
- GHS signal: GHS02
: P210