Orders LinkContactUsLinkComponent
Flat mops
Flat mops
Supplier:   MICRONOVA
Flat mops
Supplier:  MICRONOVA
Certificates
Flat mops
5 Options Available Below
Product Details & Documents
These brick-style, flat sponge mop heads are excellent for applying disinfectant in pharmaceutical production or aseptic processing applications.
  • Easily clean large surfaces
  • Heavy-duty construction offers additional resistance to textured floors and aggressive cleaning methods
  • Fully padded mop heads can be laundered and reused several times
Ideal for cleaning large surface areas as well as reaching shelving, doorjambs, and corners.
Mop heads feature heavy-duty construction with PolyTex™ textured polyester material, PolyTex™ material combined with MegaTex™ textured non-woven polyamide fabric strip, or PolyTex™ material combined with microfibre fabric strip. PolyTex™/MegaTex™ mop heads offer additional resistance to textured floors and more aggressive action for spills and heavy soils and these heads allow faster pick up of moisture in standard clean/disinfect applications such as hospitals and animal labs.
Bestellinformation: All components must be purchased separately. An electropolished stainless steel quick-disconnect adapter is required to attach mop head to an electropolished stainless steel extension handle.
Product Family Options
Product Information
  • Length
    Availability
    Price
  • MICRMZFM7-9
    PolyTex™ mop heads
    PolyTex™ mop heads
    22,86 cm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 30,00 €
    30,00 €
    1 * 1 ST
       30,00 €
  • MICRMZFM7-14
    PolyTex™ mop heads
    PolyTex™ mop heads
    35,56 cm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 36,20 €
    36,20 €
    1 * 1 ST
       36,20 €
  • MICRMZFM7-14IR
    PolyTex™ mop heads, gamma-irradiated
    PolyTex™ mop heads, gamma-irradiated
    35,56 cm
    Log in for availability
    Log in for availability
    1 * 24 ST
    1 * 24 ST - 1 090,00 €
    1 090,00 €
    1 * 24 ST
       1 090,00 €
  • MICRMZFM7-18
    PolyTex™ mop heads
    PolyTex™ mop heads
    45,72 cm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 39,70 €
    39,70 €
    1 * 1 ST
       39,70 €
  • MICRMZFM7-18IR
    PolyTex™ mop heads, gamma-irradiated
    PolyTex™ mop heads, gamma-irradiated
    45,72 cm
    Log in for availability
    Log in for availability
    1 * 24 ST
    1 * 24 ST - 1 210,00 €
    1 210,00 €
    1 * 24 ST
       1 210,00 €