Order Entry
Austria
ContactUsLinkComponent
 
Chinese ink, red, Q Path® stain for histology
Catalog # 01816204.
Supplier:  VWR Chemicals
CAS Number:  
undefined
Chinese ink, red, Q Path® stain for histology
Catalog # 01816204.
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    30 mL
  • Seq:
    2345

Specifications

Specification Test Results

  • Appearance:
    Red liquid
  • Colour of solution:
    Passes test
  • Density (20/4):
    1.130 --> 1.170

Specification Test Results

About this item

 :
 :1333-86-4