Chinese ink, blue, Q Path® stain for histology
Catalog # 01816203.
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:30 mL
- Seq:2345
Specifications
Specification Test Results
- Appearance:Blue liquid
- Colour of solution:Passes test
- Density (20/4):1.190 --> 1.370
Specification Test Results
About this item
:
:1333-86-4