Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Petroleum Industry Standards
Solvent screen, certified reference material, Supelco®
Solvent screen, certified reference material, Supelco®
Solvent screen, certified reference material, Supelco®
Catalog # SQC070-20ML
Supplier:
Merck
CAS Number:
Expand image
Solvent screen, certified reference material, Supelco®
Catalog # SQC070-20ML
Supplier:
Merck
CAS Number:
Specifications
Pk:
20 mL
Standards:
Certified reference material
Description:
Solvent Screen
Specifications
About this item
Product Details
Product Details