Order Entry
Austria
ContactUsLinkComponent
Supelco® Cerilliant® Organic Reference Standard, 1,25-Dihydroxyvitamin D3-¹³C₃ (25,26,27-¹³C₃) solution
Supelco® Cerilliant® Organic Reference Standard, 1,25-Dihydroxyvitamin D3-¹³C₃ (25,26,27-¹³C₃) solution
Catalog # H-107-1ML
Supplier:  Merck
CAS Number:  
undefined
Supelco® Cerilliant® Organic Reference Standard, 1,25-Dihydroxyvitamin D3-¹³C₃ (25,26,27-¹³C₃) solution
Catalog # H-107-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    5 µg/ml in ethanol
  • Pk:
    1 mL
  • Standards:
    Certified reference material, Cerilliant®
  • Description:
    1,25-Dihydroxyvitamin D3-¹³C₃ (25,26,27-¹³C₃) solution
  • Storage conditions:
    –70 °C
  • Hazard class description:
    H225 - H319
  • Shipping conditions:
    DICE
  • M-Clarity quality level:
    MQ300
  • Melting point:
    –143.99 °C
  • Molecular weight:
    419,61
  • Flash point:
    14,0 °C - Closed cup
  • Pk Info:
    Ampule of 1 ml

Specifications

About this item

  • GHS signal: GHS02, GHS07
Caution: P210 - P233 - P240 - P241 - P242 - P305 + P351 + P338