Order Entry
Austria
ContactUsLinkComponent
n-Paraffin Mix C10, C12, C14, C16, certified reference material, Supelco®
n-Paraffin Mix C10, C12, C14, C16, certified reference material, Supelco®
Catalog # 47102.
Supplier:  Merck
CAS Number:  
n-Paraffin Mix C10, C12, C14, C16, certified reference material, Supelco®
Catalog # 47102.
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    (varied conc.)
  • Pk:
    1
  • Description:
    n-Paraffin Mix C10, C12, C14, C16

Specifications

About this item