ContactUsLinkComponent
ASTM® D5442 quantitative linearity standard, C12 - C50, analytical standard, Supelco®
  502243.
 :  Merck
 :  
ASTM® D5442 quantitative linearity standard, C12 - C50, analytical standard, Supelco®
  502243.
 :  Merck
 :  
  • Concentration:
    0.01 % (w/w) each component in cyclohexane
  • Pk:
    1
  • Description:
    ASTM(R) D5442 Quantitative Linearity Standard, C12 - C50