Order Entry
Austria
ContactUsLinkComponent
ASTM® D5307 crude oil quantitative standard, analytical standard, Supelco®
ASTM® D5307 crude oil quantitative standard, analytical standard, Supelco®
Catalog # 48179.
Supplier:  Merck
CAS Number:  
ASTM® D5307 crude oil quantitative standard, analytical standard, Supelco®
Catalog # 48179.
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    6.25 % (w/w) each component
  • Pk:
    1
  • Description:
    ASTM(R) D5307 Crude oil quantitative standard

Specifications

About this item