Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Petroleum Industry Standards
ASTM® D4815 valve timing mix, analytical standard, Supelco®
ASTM® D4815 valve timing mix, analytical standard, Supelco®
ASTM® D4815 valve timing mix, analytical standard, Supelco®
Catalog # 47212.
Supplier:
Merck
CAS Number:
Expand image
ASTM® D4815 valve timing mix, analytical standard, Supelco®
Catalog # 47212.
Supplier:
Merck
CAS Number:
Specifications
Concentration:
(each component at the nominal wt% indicated)
Pk:
1
Description:
ASTM(R) D4815 Valve Timing Mix
Specifications
About this item
Product Details
Product Details